Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-701,324, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

142326-59-8
C21H14ClNO3
363.797
FLVRDMUHUXVRET-UHFFFAOYSA-N
7-Chloro-4-hydroxy-3-(3-phenoxy)phenyl-2(1H)-quinolinone
54682505
7-chloro-4-hydroxy-3-(3-phenoxyphenyl)-1H-quinolin-2-one
C1=CC=C(C=C1)OC2=CC=CC(=C2)C3=C(C4=C(C=C(C=C4)Cl)NC3=O)O

NMDA antagonist; acts at glycine site

An orally active and long acting anticonvulsant with high affinity and selectivity for the glycine site on the NMDA receptor.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 363.8
Chemical Name or Material L-701, 324
Grade >99%

Recommended Storage : Store at Room Temperature