Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-703,664 succinate, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
144776-01-2
C20H30N4O6S
454.542
WLXRLNXDMGIORD-UHFFFAOYSA-N
l-703,664 succinate, n,n-dimethyl-5-5-methyl-1,1-dioxodo-1,2,5-thiadiazolidin-2-yl methyl-1h-indole-3-ethanamine succinate
90488774
butanedioic acid;N,N-dimethyl-2-[5-[(5-methyl-1,1-dioxo-1,2,5-thiadiazolidin-2-yl)methyl]-1H-indol-3-yl]ethanamine
CN1CCN(S1(=O)=O)CC2=CC3=C(C=C2)NC=C3CCN(C)C.C(CC(=O)O)C(=O)O
This item is not returnable. View return policy
144776-01-2
C20H30N4O6S
454.542
WLXRLNXDMGIORD-UHFFFAOYSA-N
l-703,664 succinate, n,n-dimethyl-5-5-methyl-1,1-dioxodo-1,2,5-thiadiazolidin-2-yl methyl-1h-indole-3-ethanamine succinate
90488774
butanedioic acid;N,N-dimethyl-2-[5-[(5-methyl-1,1-dioxo-1,2,5-thiadiazolidin-2-yl)methyl]-1H-indol-3-yl]ethanamine
CN1CCN(S1(=O)=O)CC2=CC3=C(C=C2)NC=C3CCN(C)C.C(CC(=O)O)C(=O)O

Selective 5-HT1D receptor agonist

5-HT1D receptor agonist that displays selectivity over other 5-HT receptor subtypes (pIC50 values are 7.2, 6.1, < 5.0 and < 5.0 for 5-HT1D, 5-HT1A, 5-HT2A and 5-HT3 respectively) and other receptors (pIC50 < 5.0 for adenosine, adrenergic, excitatory amino acids, dopamine, histamine, muscarinic, nicotinic and opiate receptors).
Quantity 10 mg
Formula Weight 454.54
Percent Purity >99%
Chemical Name or Material L-703, 664 succinate

Recommended Storage : Store at +4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit