Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-703,664 succinate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
5-HT1D receptor agonist that displays selectivity over other 5-HT receptor subtypes (pIC50 values are 7.2, 6.1, < 5.0 and < 5.0 for 5-HT1D, 5-HT1A, 5-HT2A and 5-HT3 respectively) and other receptors (pIC50 < 5.0 for adenosine, adrenergic, excitatory amino acids, dopamine, histamine, muscarinic, nicotinic and opiate receptors).
Specifications
Quantity | 50 mg |
Formula Weight | 454.54 |
Percent Purity | >99% |
Chemical Name or Material | L-703, 664 succinate |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction