Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-732,138, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
148451-96-1
C22H18F6N2O3
472.387
BYYQYXVAWXAYQC-IBGZPJMESA-N
3,5-bis tfm bz nactrp,3,5-bis trifluoromethyl benzyl n-acetyltryptophan,n-acetyltryptophan 3,5-bis trifluoromethyl benzyl ester,chembl22870,n-acetyl-l-trp-3,5-bistrifluoromethylbenzyl ester,3,5-bis trifluoromethyl phenyl methyl 2s-2-acetamido-3-1h-indol-3-yl propanoate,n-acetyl-l-tryptophan-3,5-bistrifluoromethylbenzyl ester,n-acetyl-l-tryptophan 3,5-bis trifluoromethyl benzyl ester,l-tryptophan, n-acetyl-, 3,5-bis trifluoromethyl phenyl methyl ester,tocris-0868
132837
[3,5-bis(trifluoromethyl)phenyl]methyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
CC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)OCC3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F
148451-96-1
C22H18F6N2O3
472.387
BYYQYXVAWXAYQC-IBGZPJMESA-N
3,5-bis tfm bz nactrp,3,5-bis trifluoromethyl benzyl n-acetyltryptophan,n-acetyltryptophan 3,5-bis trifluoromethyl benzyl ester,chembl22870,n-acetyl-l-trp-3,5-bistrifluoromethylbenzyl ester,3,5-bis trifluoromethyl phenyl methyl 2s-2-acetamido-3-1h-indol-3-yl propanoate,n-acetyl-l-tryptophan-3,5-bistrifluoromethylbenzyl ester,n-acetyl-l-tryptophan 3,5-bis trifluoromethyl benzyl ester,l-tryptophan, n-acetyl-, 3,5-bis trifluoromethyl phenyl methyl ester,tocris-0868
132837
[3,5-bis(trifluoromethyl)phenyl]methyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
CC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)OCC3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F

Potent, selective NK1 antagonist

Potent and highly selective competitive tachykinin NK1 receptor antagonist (IC50 = 2.3 nM).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Formula Weight 472.39
Percent Purity >98%
Quantity 50mg
Chemical Name or Material L-732, 138

Recommended Storage : Store at Room Temperature