Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-745,870 trihydrochloride, Tocris

Highly selective D4 antagonist

Manufacturer:  R&D Systems 1002/50

Quantity 50mg

 View more versions of this product

Catalog No. 10-025-0


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

>99%
436.21
L-745, 870 trihydrochloride
C18H19N4Cl·3HCl
3-(4-[4-Chlorophenyl]piperazin-1-yl)-methyl-1H-pyrrolo[2, 3-b]pyridine trihydrochloride

Specifications

50mg
>99%
436.21
L-745, 870 trihydrochloride
866021-03-6
C18H19N4Cl·3HCl
3-(4-[4-Chlorophenyl]piperazin-1-yl)-methyl-1H-pyrrolo[2, 3-b]pyridine trihydrochloride

Safety and Handling

Recommended Storage : Desiccate at +4°C

Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit