Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-759,656, Tocris Bioscience™

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
174627-56-6
C26H40O2
384.604
BJIIKHXAZBTGLF-NHCUHLMSSA-N
6ar,10ar-1-methoxy-6,6-dimethyl-9-methylidene-3-2-methyloctan-2-yl-7,8,10,10a-tetrahydro-6ah-benzo c chromene,6ar,10ar-3-1,1-dimethylheptyl-6a,7,8,9,10,10a-hexahydro-1-methoxy-6,6-dimethyl-9-methylene-6h-dibenzo b,d pyran,d02inf,gtpl749,6ar,10ar-3-1,1-dimethyl-heptyl-1-methoxy-6,6-dimethyl-9-methylene-6a,7,8,9,10,10a-hexahydro-6h-benzo c chromene
5311216
(6aR,10aR)-1-methoxy-6,6-dimethyl-9-methylidene-3-(2-methyloctan-2-yl)-7,8,10,10a-tetrahydro-6aH-benzo[c]chromene
CCCCCCC(C)(C)C1=CC2=C(C3CC(=C)CCC3C(O2)(C)C)C(=C1)OC
This item is not returnable. View return policy
174627-56-6
C26H40O2
384.604
BJIIKHXAZBTGLF-NHCUHLMSSA-N
6ar,10ar-1-methoxy-6,6-dimethyl-9-methylidene-3-2-methyloctan-2-yl-7,8,10,10a-tetrahydro-6ah-benzo c chromene,6ar,10ar-3-1,1-dimethylheptyl-6a,7,8,9,10,10a-hexahydro-1-methoxy-6,6-dimethyl-9-methylene-6h-dibenzo b,d pyran,d02inf,gtpl749,6ar,10ar-3-1,1-dimethyl-heptyl-1-methoxy-6,6-dimethyl-9-methylene-6a,7,8,9,10,10a-hexahydro-6h-benzo c chromene
5311216
(6aR,10aR)-1-methoxy-6,6-dimethyl-9-methylidene-3-(2-methyloctan-2-yl)-7,8,10,10a-tetrahydro-6aH-benzo[c]chromene
CCCCCCC(C)(C)C1=CC2=C(C3CC(=C)CCC3C(O2)(C)C)C(=C1)OC

Highly selective CB2 agonist

Highly selective CB 2 receptor agonist (K i values are 11.8 and 4888 nM for CB 2 and CB 1 receptors respectively). Potently inhibits forskolin-stimulated cAMP production via CB 2 receptors expressed in CHO cells (EC 50 = 3.1 nM).
Quantity 10mg
Formula Weight 384.6
Percent Purity >98%
Chemical Name or Material L-759, 656

Recommended Storage : Store at -20°C