L-779,450, Tocris

Potent Raf kinase inhibitor

Manufacturer: R&D Systems 3185/10

Quantity 10mg

 View more versions of this product

Catalog No. 31-851-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C20H14ClN3O
    Formula Weight 347.8
    Synonym 2-Chloro-5-[2-Phenyl-5-(4-pyridinyl)-1H-imidazol-4-yl]phenol
    Chemical Name or Material L-779, 450

    Specifications

    Formula Weight 347.8
    Quantity 10mg
    Assay Percent Range >99%
    Molecular Formula C20H14ClN3O
    Synonym 2-Chloro-5-[2-Phenyl-5-(4-pyridinyl)-1H-imidazol-4-yl]phenol
    Recommended Storage Store at +4°C
    CAS 303727-31-3
    Chemical Name or Material L-779, 450

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit