Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

l-Afadin Mouse anti-Canine, Rat, Clone: 3, MilliporeSigma™

Mouse Monoclonal Antibody

$373.00

Specifications

Antigen l-Afadin
Clone 3
Concentration 0.5 mg/mL
Applications Immunocytochemistry, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABC1755100
View Documents
MilliporeSigma
MABC1755100UL
100 μL
Each for $373.00
Add to cart
 
MABC175525
View Documents
MilliporeSigma
MABC175525UL
25 μL
Each for $373.00
Add to cart
 
Description

Description

l-Afadin Monoclonal antibody specifically detects l-Afadin in Canine, Rat samples. It is validated for Immunocytochemistry, Western Blotting.
Specifications

Specifications

l-Afadin
0.5 mg/mL
Unconjugated
Mouse
Canine, Rat
KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal region of rat Afadin.
Monoclonal
Protein G purified
3
Immunocytochemistry, Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1κ
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit