Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alaninamide Hydrochloride 97.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
33208-99-0
C3H9ClN2O
124.568
FIAINKIUSZGVGX-DKWTVANSSA-N
l-alaninamide hydrochloride,l-alaninamide hcl,s-2-aminopropanamide hydrochloride,l-alanine amide hydrochloride,2s-2-aminopropanamide hydrochloride,s-2-aminopropionamide hydrochloride,h-ala-nh2.hcl,ala-nh2 hcl,alaninamide, hydrochloride,l-alaninamidehydrochloride
2775816
(2S)-2-aminopropanamide;hydrochloride
CC(C(=O)N)N.Cl
33208-99-0
C3H9ClN2O
124.568
FIAINKIUSZGVGX-DKWTVANSSA-N
l-alaninamide hydrochloride,l-alaninamide hcl,s-2-aminopropanamide hydrochloride,l-alanine amide hydrochloride,2s-2-aminopropanamide hydrochloride,s-2-aminopropionamide hydrochloride,h-ala-nh2.hcl,ala-nh2 hcl,alaninamide, hydrochloride,l-alaninamidehydrochloride
2775816
(2S)-2-aminopropanamide;hydrochloride
CC(C(=O)N)N.Cl
Melting Point 217°C
Quantity 25g
Color White
Formula Weight 124.57
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (N)
Chemical Name or Material L-Alaninamide Hydrochloride