Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Alanine benzyl ester hydrochloride, 98%

Click to view available options
Quantity:
5g
25g
100g
5557-83-5
C10H14ClNO2
215.677
MFCD00054340
RLMHWGDKMJIEHH-QRPNPIFTSA-N
h-ala-obzl.hcl,l-alanine benzyl ester hydrochloride,h-ala-obzl hcl,s-benzyl 2-aminopropanoate hydrochloride,l-alanine benzyl ester hcl,benzyl l-alaninate hydrochloride,benzyl 2s-2-aminopropanoate hydrochloride,h-ala-obzlhcl,ala-obzl hcl,nh2-ala-obn hcl
12210893
benzyl (2S)-2-aminopropanoate;hydrochloride
CC(C(=O)OCC1=CC=CC=C1)N.Cl
5557-83-5
C10H14ClNO2
215.677
MFCD00054340
RLMHWGDKMJIEHH-QRPNPIFTSA-N
h-ala-obzl.hcl,l-alanine benzyl ester hydrochloride,h-ala-obzl hcl,s-benzyl 2-aminopropanoate hydrochloride,l-alanine benzyl ester hcl,benzyl l-alaninate hydrochloride,benzyl 2s-2-aminopropanoate hydrochloride,h-ala-obzlhcl,ala-obzl hcl,nh2-ala-obn hcl
12210893
benzyl (2S)-2-aminopropanoate;hydrochloride
CC(C(=O)OCC1=CC=CC=C1)N.Cl
Formula Weight 215.68
Physical Form Powder
Percent Purity 98%
Color White
Quantity 5g
Chemical Name or Material L-Alanine benzyl ester hydrochloride

storageNote1 : -30° to -10°C