Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Alanine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
L-Alanine methyl ester hydrochloride is used to make in-vivo measurement of glucose and alanine metabolism in studies of patients with diabetes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Solubility
Soluble in Water (100 mg/ml).
Notes
Hygroscopic store under inert gas Store away from strong oxidizing agents.
Solubility
Soluble in Water (100 mg/ml).
Notes
Hygroscopic store under inert gas Store away from strong oxidizing agents.
Specifications
Melting Point | 106°C to 110°C |
Beilstein | 3594033 |
Solubility Information | Soluble in Water (100mg/ml). |
Formula Weight | 139.58 |
Percent Purity | 99% |
Quantity | 25 g |
Optical Rotation | +6.5° (c=10 in methanol) |
Sensitivity | Hygroscopic |
Chemical Name or Material | L-Alanine methyl ester hydrochloride |
Safety and Handling
EINECSNumber | 219-652-4 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction