Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alanine, Tocris Bioscience™

Catalog Number 02051g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
56-41-7
C3H7NO2
89.09
MFCD00064410
QNAYBMKLOCPYGJ-REOHCLBHSA-N
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
5950
CHEBI:16977
(2S)-2-aminopropanoic acid
C[C@H](N)C(O)=O
This item is not returnable. View return policy
56-41-7
C3H7NO2
89.09
MFCD00064410
QNAYBMKLOCPYGJ-REOHCLBHSA-N
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
5950
CHEBI:16977
(2S)-2-aminopropanoic acid
C[C@H](N)C(O)=O

Glycine receptor agonist

Agonist at the inhibitory glycine receptor.
Formula Weight 89.09
Percent Purity >99%
Quantity 1 g
Chemical Name or Material L-Alanine
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit