Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Alanyl-L-glutamine, 99%

Click to view available options
Quantity:
5g
25g
100g
39537-23-0
C8H15N3O4
217.225
MFCD00133046
HJCMDXDYPOUFDY-WHFBIAKZSA-N
l-alanyl-l-glutamine,ala-gln,alanyl-glutamine,alanyl glutamine,alanylglutamine,l-glutamine, l-alanyl,dipeptamin,h-ala-gln-oh,n 2-l-alanyl-l-glutamine,glutamax
123935
CHEBI:73788
(2S)-5-amino-2-[[(2S)-2-aminopropanoyl]amino]-5-oxopentanoic acid
CC(C(=O)NC(CCC(=O)N)C(=O)O)N
39537-23-0
C8H15N3O4
217.225
MFCD00133046
HJCMDXDYPOUFDY-WHFBIAKZSA-N
l-alanyl-l-glutamine,ala-gln,alanyl-glutamine,alanyl glutamine,alanylglutamine,l-glutamine, l-alanyl,dipeptamin,h-ala-gln-oh,n 2-l-alanyl-l-glutamine,glutamax
123935
CHEBI:73788
(2S)-5-amino-2-[[(2S)-2-aminopropanoyl]amino]-5-oxopentanoic acid
CC(C(=O)NC(CCC(=O)N)C(=O)O)N
Solubility
Soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.Store in a cool place. Incompatible with strong oxidizing agents.
Solubility Information Soluble in water
Formula Weight 217.22
Physical Form Powder
Percent Purity 99%
Boiling Point 615°C
Color White
Melting Point 222°C
Quantity 5g
Chemical Name or Material L-Alanyl-L-glutamine

RTECSNumber : MA2275262

storageNote1 : 2° to 8°C