Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Alanyl-L-leucine, 95%

Click to view available options
Quantity:
250mg
1g
5g
3303-34-2
C9H18N2O3
202.254
MFCD00065106
RDIKFPRVLJLMER-BQBZGAKWSA-N
ala-leu,l-alanyl-l-leucine,n-l-alanyl-l-leucine,h-ala-leu-oh,alanyl-dl-leucine,2s-2-2s-2-aminopropanamido-4-methylpentanoic acid,leucine, alanyl,l-ala-l-leu,l-ala-l-leu-oh,l-leucine, l-alanyl
96801
CHEBI:73770
(2S)-2-[[(2S)-2-aminopropanoyl]amino]-4-methylpentanoic acid
CC(C)CC(C(=O)O)NC(=O)C(C)N
3303-34-2
C9H18N2O3
202.254
MFCD00065106
RDIKFPRVLJLMER-BQBZGAKWSA-N
ala-leu,l-alanyl-l-leucine,n-l-alanyl-l-leucine,h-ala-leu-oh,alanyl-dl-leucine,2s-2-2s-2-aminopropanamido-4-methylpentanoic acid,leucine, alanyl,l-ala-l-leu,l-ala-l-leu-oh,l-leucine, l-alanyl
96801
CHEBI:73770
(2S)-2-[[(2S)-2-aminopropanoyl]amino]-4-methylpentanoic acid
CC(C)CC(C(=O)O)NC(=O)C(C)N
Beilstein 1726651
Formula Weight 202.25
Physical Form Solid
Percent Purity 95%
Quantity 1g
Chemical Name or Material L-Alanyl-L-leucine

storageNote1 : -30° to -10°C