Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-allo-Threonine, 99%

Manufacturer:  Alfa Aesar™ H27301MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27301MD


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

28954-12-3
119.12
1721645
L-allo-Threonine
C4H9NO3
MFCD00064268
(2S,3S)-2-Amino-3-hydroxybutyric acid; H-allo-Thr-OH

Specifications

28954-12-3
119.12
1721645
272°C
+9.2° (c=2 in Water)
99%
C4H9NO3
MFCD00064268
(2S,3S)-2-Amino-3-hydroxybutyric acid; H-allo-Thr-OH
250mg
L-allo-Threonine

Safety and Handling

SDS & Safety Documents

RTECSNumber : BA4055000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit