Alfa Aesar™ L-allo-Threonine, 99%

Manufacturer: Alfa Aesar™ H2730103

Quantity 1g

 View more versions of this product

Catalog No. AAH2730103

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 28954-12-3
    Molecular Formula C4H9NO3
    Formula Weight 119.12
    MDL Number MFCD00064268
    Beilstein 1721645
    Synonym (2S,3S)-2-Amino-3-hydroxybutyric acid; H-allo-Thr-OH
    Chemical Name or Material L-allo-Threonine

    Specifications

    Formula Weight 119.12
    MDL Number MFCD00064268
    CAS 28954-12-3
    Synonym (2S,3S)-2-Amino-3-hydroxybutyric acid; H-allo-Thr-OH
    Melting Point 272°C
    Quantity 1g
    Optical Rotation +9.2° (c=2 in Water)
    Molecular Formula C4H9NO3
    Beilstein 1721645
    RTECS Number BA4055000
    Chemical Name or Material L-allo-Threonine
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit