Alfa Aesar™ L-(-)-alpha-Amino-gamma-butyrolactone hydrobromide, 98%

Manufacturer: Alfa Aesar™ B2314803

Quantity 1g

 View more versions of this product

Catalog No. AAB2314803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15295-77-9
    Molecular Formula C4H7NO2·HBr
    Formula Weight 182.02
    MDL Number MFCD00012723
    Beilstein 3912375
    Chemical Name or Material L-(-)-alpha-Amino-gamma-butyrolactone hydrobromide

    Specifications

    Formula Weight 182.02
    Melting Point 258° to 260°C
    Quantity 1g
    Optical Rotation −21° (c=1 in Water)
    Molecular Formula C4H7NO2·HBr
    MDL Number MFCD00012723
    CAS 15295-77-9
    Beilstein 3912375
    Chemical Name or Material L-(-)-alpha-Amino-gamma-butyrolactone hydrobromide
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit