Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-(-)-alpha-Amino-gamma-butyrolactone hydrobromide, 98%

Click to view available options
Quantity:
0.25g
1g
15295-77-9
C4H8BrNO2
182.017
MFCD00012723
MKLNTBLOABOJFZ-DFWYDOINSA-N
s-3-aminodihydrofuran-2 3h-one hydrobromide,s---alpha-amino-gamma-butyrolactone hydrobromide,l-homoserine lactone hydrobromide,s-2-amino-4-butyrolactone hydrobromide,l---alpha-amino-gamma-butyrolactone hydrobromide,2 3h-furanone, 3-aminodihydro-, hydrobromide, 3s,s---alpha-amino-gamma-butyrolactonehydrobromide,ksc924k2j,l---?-amino-?-butyrolactone hydrobromide,s---alpha-amino-gamma-butyrolactone hbr
16213138
(3S)-3-aminooxolan-2-one;hydrobromide
C1COC(=O)C1N.Br
15295-77-9
C4H8BrNO2
182.017
MFCD00012723
MKLNTBLOABOJFZ-DFWYDOINSA-N
s-3-aminodihydrofuran-2 3h-one hydrobromide,s---alpha-amino-gamma-butyrolactone hydrobromide,l-homoserine lactone hydrobromide,s-2-amino-4-butyrolactone hydrobromide,l---alpha-amino-gamma-butyrolactone hydrobromide,2 3h-furanone, 3-aminodihydro-, hydrobromide, 3s,s---alpha-amino-gamma-butyrolactonehydrobromide,ksc924k2j,l---?-amino-?-butyrolactone hydrobromide,s---alpha-amino-gamma-butyrolactone hbr
16213138
(3S)-3-aminooxolan-2-one;hydrobromide
C1COC(=O)C1N.Br
Quantity 0.25g
Optical Rotation −21° (c=1 in Water)
Melting Point 258°C to 260°C
Beilstein 3912375
Formula Weight 182.02
Percent Purity 98%
Chemical Name or Material L-(-)-alpha-Amino-gamma-butyrolactone hydrobromide