Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-alpha-Amino-N-Butyric acid, >98%, Affymetrix/USB™

Click to view available options
Quantity:
1g
5g
Packaging:
Plastic bottle
1492-24-6
C4H9NO2
103.121
MFCD00064415
QWCKQJZIFLGMSD-VKHMYHEASA-N
l-2-aminobutyric acid,s-2-aminobutanoic acid,2s-2-aminobutanoic acid,h-abu-oh,l-alpha-aminobutyric acid,s-+-2-aminobutyric acid,l-+-2-aminobutyric acid,--2-aminobutyric acid,s-2-aminobutyric acid,s-2-amino-butyric acid
80283
CHEBI:35619
(2S)-2-aminobutanoic acid
CCC(C(=O)O)N
1492-24-6
C4H9NO2
103.121
MFCD00064415
QWCKQJZIFLGMSD-VKHMYHEASA-N
l-2-aminobutyric acid,s-2-aminobutanoic acid,2s-2-aminobutanoic acid,h-abu-oh,l-alpha-aminobutyric acid,s-+-2-aminobutyric acid,l-+-2-aminobutyric acid,--2-aminobutyric acid,s-2-aminobutyric acid,s-2-amino-butyric acid
80283
CHEBI:35619
(2S)-2-aminobutanoic acid
CCC(C(=O)O)N
Packaging Plastic bottle
Melting Point >300°C
Quantity 5g
Linear Formula CH3CH2CH(NH2)COOH
Formula Weight 103.12
Physical Form Crystalline Powder
Percent Purity ≥98%
Chemical Name or Material L-alpha-Amino-N-Butyric acid