Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(-)-Altrose, 99%, ACROS Organics™

Click to view available options
Quantity:
50mg
Packaging:
Glass bottle
1949-88-8
C6H12O6
180.156
MFCD00792525
GZCGUPFRVQAUEE-AZGQCCRYSA-N
2r,3s,4s,5s-2,3,4,5,6-pentahydroxyhexanal,l-altrose hplc
102190
(2R,3S,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
C(C(C(C(C(C=O)O)O)O)O)O
1949-88-8
C6H12O6
180.156
MFCD00792525
GZCGUPFRVQAUEE-AZGQCCRYSA-N
2r,3s,4s,5s-2,3,4,5,6-pentahydroxyhexanal,l-altrose hplc
102190
(2R,3S,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
C(C(C(C(C(C=O)O)O)O)O)O
Packaging Glass bottle
Color White
Sulfated Ash 0.05% max.
Infrared Spectrum Authentic
Quantity 50mg
Specific Rotation -30° to -34° (20°C, 589nm) (c=4, H2O)
Assay Percent Range 99%
Formula Weight 180.16
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material L(-)-Altrose, 99%
Show More Show Less