Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L(+)-Amethopterin hydrate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• L(+)-Amethopterin hydrate, is a potent inhibitor of dihydrofolate reductase
• It has tumoricidal activity
Applications
• L(+)-Amethopterin hydrate is an inhibitor of dihydrofolate reductase
• It can be used in anti-tumor studies
• It is a cell cycle arrest agent, which can inhibit synthesis of DNA, RNA, and proteins
• It can also inhibit production of pro-inflammatory cytokines
Specifications
Melting Point | 195°C |
Color | Yellow to Orange |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (HPLC) |
Packaging | Glass Bottle |
Quantity | 25 mg |
Merck Index | 14, 5985 |
Specific Rotation | + 21.5 |
Solubility Information | Solubility in water: insoluble in water. Other solubilities: insoluble in alcohol,chloroform,ether,slightly soluble in dilute hydrochloric acid,soluble in dilute solutions of alkali hydroxides a |
Formula Weight | 454.44 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement: Toxic if swallowed. May cause genetic defects. May damage the unborn child. |
Precautionary Statement | GHS P Statement: IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction