Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(+)-Amethopterin hydrate, 99%, ACROS Organics™

Click to view available options
Quantity:
25mg
100mg
500mg
Packaging:
Glass Bottle
133073-73-1
C20H22N8O5·xH2O
454.44
MFCD00150847
TXQDMGIRZSHAQM-GXKRWWSZSA-N
l +-amethopterin dihydrate,methotrexate dihydrate,dihydrate methotrexate,2s-2-4-2,4-diaminopteridin-6-yl methyl methylamino phenyl carbonylamino pentanedioic acid, hydrate, hydrate
45157423
(2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid;dihydrate
CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O.O.O
133073-73-1
C20H22N8O5·xH2O
454.44
MFCD00150847
TXQDMGIRZSHAQM-GXKRWWSZSA-N
l +-amethopterin dihydrate,methotrexate dihydrate,dihydrate methotrexate,2s-2-4-2,4-diaminopteridin-6-yl methyl methylamino phenyl carbonylamino pentanedioic acid, hydrate, hydrate
45157423
(2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid;dihydrate
CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O.O.O
Melting Point 195°C
Packaging Glass Bottle
Specific Rotation + 21.5
Specific Rotation Condition + 21.50 (20.00°C c=1,14g/l Na2CO3)
Quantity 500mg
Water 13% max. (K.F.)
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Merck Index 14, 5985
Solubility Information Solubility in water: insoluble in water. Other solubilities: insoluble in alcohol,chloroform,ether,slightly soluble in dilute hydrochloric acid,soluble in dilute solutions of alkali hydroxides a
Formula Weight 454.44
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material L(+)-Amethopterin hydrate
Show More Show Less

GHS H Statement:
Toxic if swallowed.
May cause genetic defects.
May damage the unborn child.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger