Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ L-Amino Acid Oxidase (C. adamanteus)
Used for Enzyme
Supplier: Abnova™ P5236
Description
#REF!Specifications
Enzyme Activity | |
L-Amino Acid Oxidase (C. adamanteus) | |
2 mg | |
RUO | |
L-Amino Acid Oxidase Assay | |
Snake | |
Native protein purified from Crotalus adamanteus Venom |
140kDa | |
Native protein purified from Crotalus adamanteus Venom | |
Store at 4°C. Do not freeze. This product is stable for at least 3 months. | |
L-AMINO | |
≥4 units per mg protein. One Unit oxidizes one micormole of L-leucine per minute at 25°C, pH 7.6. | |
None | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction