Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-AP4, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
1mg
10mg
50mg
23052-81-5
C4H10NO5P
183.1
DDOQBQRIEWHWBT-VKHMYHEASA-N
l-ap4,2s-2-amino-4-phosphonobutanoic acid,l-+-2-amino-4-phosphonobutyric acid,chembl33567,s-2-amino-4-phosphonobutyrate,l-ap-4,3h ap4,l-2-amino-4-phosphonobutiric acid,l-1-amino-4-phosphonobutanoic acid,s-2-amino-4-phosphonobutanoic acid
179394
(2S)-2-amino-4-phosphonobutanoic acid
C(CP(=O)(O)O)C(C(=O)O)N
23052-81-5
C4H10NO5P
183.1
DDOQBQRIEWHWBT-VKHMYHEASA-N
l-ap4,2s-2-amino-4-phosphonobutanoic acid,l-+-2-amino-4-phosphonobutyric acid,chembl33567,s-2-amino-4-phosphonobutyrate,l-ap-4,3h ap4,l-2-amino-4-phosphonobutiric acid,l-1-amino-4-phosphonobutanoic acid,s-2-amino-4-phosphonobutanoic acid
179394
(2S)-2-amino-4-phosphonobutanoic acid
C(CP(=O)(O)O)C(C(=O)O)N

Selective group III mGlu agonist

Selective group III metabotropic glutamate receptor agonist. Synaptic depressant. Agonist at the quisqualate-sensitized AP6 site in hippocampus. Available as part of the Group III mGlu Receptor Tocriset and Mixed mGlu Receptor Tocriset . DL Mixture and D-isomer also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 1mg
Formula Weight 183.1
Percent Purity >99%
Chemical Name or Material L-AP4

Recommended Storage : Store at Room Temperature