Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-AP5, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

79055-67-7
C5H12NO5P
197.127
VOROEQBFPPIACJ-BYPYZUCNSA-N
L-(+)-2-Amino-5-phosphonopentanoic acid
1617110

Potent, selective NMDA antagonist. Less active form of DL-AP5 (Cat. No. 0105)

NMDA antagonist. Also agonist at quisqualate-sensitized AP6 site, where it is more potent than the isomer D-AP5 (Cat. No. 0106). Mixture DL-AP5 (Cat. No. 3693) also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
SMILES C(CC(C(=O)O)N)CP(=O)(O)O
IUPAC Name (2S)-2-amino-5-phosphonopentanoic acid
Formula Weight 197.13
Chemical Name or Material L-AP5
Grade >99%

Recommended Storage : Store at Room Temperature