Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Arginine 4-nitroanilide dihydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
5g
40127-11-5
C12H20Cl2N6O3
367.231
MFCD00058249
FBVMDLFQVOFFHS-XRIOVQLTSA-N
h-arg-pna.2hcl,s-2-amino-5-guanidino-n-4-nitrophenyl pentanamide dihydrochloride,l-arginine p-nitroanilide dihydrochloride,arginine p-nitroanilide dihydrochloride,2s-2-amino-5-carbamimidamido-n-4-nitrophenyl pentanamide dihydrochloride,h-arg-pna 2hcl,h-arg-pna.dihydrochloride,h-arg-pna inverted exclamation mark currency2hcl,n-4-nitrophenyl argininamide dihydrochloride,s-2-amino-5-guanidino-n-4-nitrophenyl pentanamide 2hcl
22811858
(2S)-2-amino-5-(diaminomethylideneamino)-N-(4-nitrophenyl)pentanamide;dihydrochloride
C1=CC(=CC=C1NC(=O)C(CCCN=C(N)N)N)[N+](=O)[O-].Cl.Cl
40127-11-5
C12H20Cl2N6O3
367.231
MFCD00058249
FBVMDLFQVOFFHS-XRIOVQLTSA-N
h-arg-pna.2hcl,s-2-amino-5-guanidino-n-4-nitrophenyl pentanamide dihydrochloride,l-arginine p-nitroanilide dihydrochloride,arginine p-nitroanilide dihydrochloride,2s-2-amino-5-carbamimidamido-n-4-nitrophenyl pentanamide dihydrochloride,h-arg-pna 2hcl,h-arg-pna.dihydrochloride,h-arg-pna inverted exclamation mark currency2hcl,n-4-nitrophenyl argininamide dihydrochloride,s-2-amino-5-guanidino-n-4-nitrophenyl pentanamide 2hcl
22811858
(2S)-2-amino-5-(diaminomethylideneamino)-N-(4-nitrophenyl)pentanamide;dihydrochloride
C1=CC(=CC=C1NC(=O)C(CCCN=C(N)N)N)[N+](=O)[O-].Cl.Cl
Formula Weight 367.23
Physical Form Solid
Percent Purity 95%
Quantity 250mg
Chemical Name or Material L-Arginine 4-nitroanilide dihydrochloride

storageNote1 : 2° to 8°C