Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Arginine 7-amido-4-methylcoumarin dihydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
50mg
Packaging:
Glass Bottle
113712-08-6
C16H23Cl2N5O3
404.292
MFCD00155578
QMGJRBUFCDSWOX-LTCKWSDVSA-N
l-arginine 7-amido-4-methylcoumarin dihydrochloride,2s-2-amino-5-carbamimidamido-n-4-methyl-2-oxochromen-7-yl pentanamide dihydrochloride,2s-2-amino-5-diaminomethylideneamino-n-4-methyl-2-oxochromen-7-yl pentanamide dihydrochloride,2s-5-amidinoamino-2-amino-n-4-methyl-2-oxochromen-7-yl pentanamide, chlor ide, chloride
2733800
(2S)-2-amino-5-(diaminomethylideneamino)-N-(4-methyl-2-oxochromen-7-yl)pentanamide;dihydrochloride
CC1=CC(=O)OC2=C1C=CC(=C2)NC(=O)C(CCCN=C(N)N)N.Cl.Cl
113712-08-6
C16H23Cl2N5O3
404.292
MFCD00155578
QMGJRBUFCDSWOX-LTCKWSDVSA-N
l-arginine 7-amido-4-methylcoumarin dihydrochloride,2s-2-amino-5-carbamimidamido-n-4-methyl-2-oxochromen-7-yl pentanamide dihydrochloride,2s-2-amino-5-diaminomethylideneamino-n-4-methyl-2-oxochromen-7-yl pentanamide dihydrochloride,2s-5-amidinoamino-2-amino-n-4-methyl-2-oxochromen-7-yl pentanamide, chlor ide, chloride
2733800
(2S)-2-amino-5-(diaminomethylideneamino)-N-(4-methyl-2-oxochromen-7-yl)pentanamide;dihydrochloride
CC1=CC(=O)OC2=C1C=CC(=C2)NC(=O)C(CCCN=C(N)N)N.Cl.Cl
Packaging Glass Bottle
Quantity 50mg
Water 3.5% max. (K.F.)
Specific Rotation Condition + 88.00 (20.00°C c=1,1N HCl)
Infrared Spectrum Authentic
Specific Rotation +86° to +90° (20°C, 589nm) (c=1, 1N HCl)
Appearance (2.5% soln. in acetic acid 80%): clear, colorless
Color White to Brown
Assay Percent Range 98%
Formula Weight 404.3
Physical Form Crystalline Powder
Percent Purity ≥97.5% (on dry substance)
Chemical Name or Material L-Arginine 7-amido-4-methylcoumarin dihydrochloride, 98%
Show More Show Less

Recommended Storage : Freezer -20°C