Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Arginine hydrochloride, 98.5 to 101.5%, Affymetrix/USB™

Click to view available options
Quantity:
500g
1kg
Packaging:
Plastic bottle
1119-34-2
C6H15ClN4O2
210.662
MFCD00064550
KWTQSFXGGICVPE-WCCKRBBISA-N
l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
66250
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
C(CC(C(=O)O)N)CN=C(N)N.Cl
1119-34-2
C6H15ClN4O2
210.662
MFCD00064550
KWTQSFXGGICVPE-WCCKRBBISA-N
l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
66250
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
C(CC(C(=O)O)N)CN=C(N)N.Cl
Linear Formula NH2C(NH)NH(CH2)3CH(NH2)COOH.HCl
Beilstein 3631658
Packaging Plastic bottle
Formula Weight 210.66
Physical Form Crystalline Powder
Percent Purity 98.5 to 101.5%
Density 1.42
Melting Point 216°C (decomposition)
Quantity 500g
Specific Gravity 1.42
Chemical Name or Material L-Arginine hydrochloride
Show More Show Less

EINECSNumber : 214-275-1

RTECSNumber : CF1995500

TSCA : Yes

Recommended Storage : Ambient temperatures