Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-arginine methyl ester dihydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
26340-89-6
C7H18Cl2N4O2
261.147
MFCD00038948
XQYZOBNLCUAXLF-XRIGFGBMSA-N
l-arginine methyl ester dihydrochloride,h-arg-ome.2hcl,methyl l-argininate dihydrochloride,h-arg-ome 2hcl,l-arginine, methyl ester, dihydrochloride,s-methyl 2-amino-5-guanidinopentanoate dihydrochloride,methyl 2s-2-amino-5-carbamimidamidopentanoate dihydrochloride,l-arginine, methyl ester, hydrochloride 1:2,h-arg-ome?hcl,h-arg-ome 2 hcl
117776
methyl (2S)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride
COC(=O)C(CCCN=C(N)N)N.Cl.Cl
26340-89-6
C7H18Cl2N4O2
261.147
MFCD00038948
XQYZOBNLCUAXLF-XRIGFGBMSA-N
l-arginine methyl ester dihydrochloride,h-arg-ome.2hcl,methyl l-argininate dihydrochloride,h-arg-ome 2hcl,l-arginine, methyl ester, dihydrochloride,s-methyl 2-amino-5-guanidinopentanoate dihydrochloride,methyl 2s-2-amino-5-carbamimidamidopentanoate dihydrochloride,l-arginine, methyl ester, hydrochloride 1:2,h-arg-ome?hcl,h-arg-ome 2 hcl
117776
methyl (2S)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride
COC(=O)C(CCCN=C(N)N)N.Cl.Cl
Assay Percent Range 98%
Linear Formula NH2C(NH)N(H)(CH2)3CH(NH2)C(O)OCH3·2HCl
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 261.15
Specific Rotation +18° to +22° (20°C, 589nm) (c=2.5, CH3OH)
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Specific Rotation Condition +20° (20°C c=2.5 Chloroform)
Color White
Quantity 5g
Chemical Name or Material L-Arginine methyl ester dihydrochloride
Show More Show Less