Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Arginine Monohydrochloride, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™

Click to view available options
Quantity:
100g
1kg
Packaging:
Poly Bottle

Chemical Identifiers

1119-34-2
C6H15ClN4O2
210.662
KWTQSFXGGICVPE-WCCKRBBISA-N
66250
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
C(CC(C(=O)O)N)CN=C(N)N.Cl

High quality biochemicals for research use

Packaging Poly Bottle
Formula Weight 210.67
Quantity 1kg
Specific Gravity 1L = 1.42kg
Chemical Name or Material L-(+)-Arginine Monohydrochloride
Grade BAKER ANALYZED™ Biochemical Reagent