Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Arginine Monohydrochloride, FCC, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
1119-34-2
C6H15ClN4O2
210.66
KWTQSFXGGICVPE-WCCKRBBISA-N
hydrogen (2S)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride
[H+].[Cl-].N[C@@H](CCCN=C(N)N)C(O)=O
1119-34-2
C6H15ClN4O2
210.66
KWTQSFXGGICVPE-WCCKRBBISA-N
hydrogen (2S)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride
[H+].[Cl-].N[C@@H](CCCN=C(N)N)C(O)=O

A1338, 1119-34-2, C6H14N4O2•HCl

Spectrum™ L-Arginine Monohydrochloride, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Ignition Residue 0.001
Lead (Pb) 5 mg/kg
Loss on Drying 0.003
Packaging Fiber Drum
Specific Rotation +21.3° to +23.5°
Percent Purity 98.5 to 101.5%
Quantity 50 kg