Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Ascorbic Acid, Powder, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™

Click to view available options
Quantity:
100g
500g
Packaging:
Poly Bottle

Chemical Identifiers

50-81-7
C6H8O6
176.124
CIWBSHSKHKDKBQ-JLAZNSOCSA-N
54670067
CHEBI:29073

High quality biochemicals for research use

CAS 50-81-7
Molecular Formula C6H8O6
Packaging Poly Bottle
InChI Key CIWBSHSKHKDKBQ-JLAZNSOCSA-N
SMILES C(C(C1C(=C(C(=O)O1)O)O)O)O
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
Molecular Weight (g/mol) 176.124
PubChem CID 54670067
ChEBI CHEBI:29073
Formula Weight 176.12
Physical Form Powder
Quantity 100g
Specific Gravity 1L = 1.65kg
Chemical Name or Material L-(+)-Ascorbic Acid
Grade BAKER ANALYZED™ Biochemical Reagent
Show More Show Less