Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Asparagine, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic Bottle
70-47-3
C4H8N2O3
132.119
MFCD00064401
DCXYFEDJOCDNAF-REOHCLBHSA-N
l-asparagine,asparagine,s-asparagine,aspartamic acid,agedoite,altheine,h-asn-oh,crystal vi,asparamide,asparagine acid
6267
CHEBI:17196
(2S)-2,4-diamino-4-oxobutanoic acid
C(C(C(=O)O)N)C(=O)N
70-47-3
C4H8N2O3
132.119
MFCD00064401
DCXYFEDJOCDNAF-REOHCLBHSA-N
l-asparagine,asparagine,s-asparagine,aspartamic acid,agedoite,altheine,h-asn-oh,crystal vi,asparamide,asparagine acid
6267
CHEBI:17196
(2S)-2,4-diamino-4-oxobutanoic acid
C(C(C(=O)O)N)C(=O)N
Assay Percent Range 99%
Linear Formula H2NCOCH2CH(NH2)CO2H
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 132.12
Specific Rotation + 34.50
Physical Form Crystalline Powder
Solubility Solubility in water: 20g/l (20°C). Other solubilities: insoluble in organic solvents
Percent Purity 99%
Water 1% max.
Color White
Quantity 500g
Chemical Name or Material L-Asparagine
Show More Show Less