Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Asparagine, Anhydrous ≥98.5%, Ultrapure, Affymetrix/USB™

Click to view available options
Quantity:
100g
1kg
Packaging:
Plastic bottle
70-47-3
C4H8N2O3
132.119
MFCD00064401
DCXYFEDJOCDNAF-REOHCLBHSA-N
l-asparagine,asparagine,s-asparagine,aspartamic acid,agedoite,altheine,h-asn-oh,crystal vi,asparamide,asparagine acid
6267
CHEBI:17196
(2S)-2,4-diamino-4-oxobutanoic acid
C(C(C(=O)O)N)C(=O)N
70-47-3
C4H8N2O3
132.119
MFCD00064401
DCXYFEDJOCDNAF-REOHCLBHSA-N
l-asparagine,asparagine,s-asparagine,aspartamic acid,agedoite,altheine,h-asn-oh,crystal vi,asparamide,asparagine acid
6267
CHEBI:17196
(2S)-2,4-diamino-4-oxobutanoic acid
C(C(C(=O)O)N)C(=O)N
Linear Formula NH2COCH2CH(NH2)COOH
Packaging Plastic bottle
Formula Weight 132.12
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Melting Point 234°C to 236°C (decomposition)
Quantity 1kg
Chemical Name or Material L-Asparagine