L-(+)-Asparagine hydrate, 98%, Alfa Aesar™

Manufacturer: Alfa Aesar 3651222

Quantity 100g

 View more versions of this product

Catalog No. AA3651222

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 5794-13-8
    Molecular Formula H2NCOCH2CH(NH2)CO2H·H2O
    Formula Weight 150.14
    MDL Number MFCD00151038
    Merck Index 14,837
    Chemical Name or Material L-(+)-Asparagine hydrate

    Specifications

    Formula Weight 150.14
    Melting Point 233° to 235°C
    Quantity 100g
    TSCA TSCA
    Physical Form Powder
    Molecular Formula H2NCOCH2CH(NH2)CO2H·H2O
    MDL Number MFCD00151038
    CAS 5794-13-8
    Merck Index 14,837
    Chemical Name or Material L-(+)-Asparagine hydrate
    Assay 98%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit