Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Asparagine Monohydrate ≥98%, Ultrapure, Affymetrix/USB™

Click to view available options
Quantity:
100g
1kg
Packaging:
Plastic bottle
5794-13-8
C4H10N2O4
150.134
MFCD00151038
RBMGJIZCEWRQES-DKWTVANSSA-N
l-asparagine monohydrate,l +-asparagine monohydrate,l-asparagine hydrate,asparagine monohydrate,h-asn-oh.h2o,l-asparagine, monohydrate,asparagine nf,s-2,4-diamino-4-oxobutanoic acid hydrate,asparagine,unii-2pd79vf521
170358
(2S)-2,4-diamino-4-oxobutanoic acid;hydrate
C(C(C(=O)O)N)C(=O)N.O
5794-13-8
C4H10N2O4
150.134
MFCD00151038
RBMGJIZCEWRQES-DKWTVANSSA-N
l-asparagine monohydrate,l +-asparagine monohydrate,l-asparagine hydrate,asparagine monohydrate,h-asn-oh.h2o,l-asparagine, monohydrate,asparagine nf,s-2,4-diamino-4-oxobutanoic acid hydrate,asparagine,unii-2pd79vf521
170358
(2S)-2,4-diamino-4-oxobutanoic acid;hydrate
C(C(C(=O)O)N)C(=O)N.O
Packaging Plastic bottle
Formula Weight 150.13
Physical Form Crystalline Powder
Percent Purity ≥98%
Melting Point 215°C to 217°C
Quantity 100g
Chemical Name or Material L-Asparagine monohydrate