Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Asparagine, Monohydrate, FCC, 98-101.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
5794-13-8
C4H10N2O4
150.13
RBMGJIZCEWRQES-DKWTVANSSA-N
(2S)-2-amino-3-carbamoylpropanoic acid hydrate
O.N[C@@H](CC(N)=O)C(O)=O
5794-13-8
C4H10N2O4
150.13
RBMGJIZCEWRQES-DKWTVANSSA-N
(2S)-2-amino-3-carbamoylpropanoic acid hydrate
O.N[C@@H](CC(N)=O)C(O)=O

A1373, 5794-13-8, C4H8N2O3•H2O

Spectrum™ L-Asparagine, Monohydrate, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.001
Optical Rotation +33.0° to +36.5°
Quantity 100 g
Loss on Drying 11.5-12.5%
CAS Max % 1
Percent Purity 98 to 101.5%