Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Asparagine tert-butyl ester hydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
5g
63094-81-5
C8H17ClN2O3
224.685
MFCD00039079
RXNKCUXXNGWROA-JEDNCBNOSA-N
h-asn-otbu hcl,l-asparagine tert-butyl ester hydrochloride,h-asn-otbu.hcl,tert-butyl l-asparaginate-hydrogen chloride 1/1,l-asparagine, 1,1-dimethylethyl ester, monohydrochloride,tert-butyl 2s-2-amino-3-carbamoylpropanoate hydrochloride,h-asn-otbucl,h-asn-otbu?cl,h-asn-otbuhcl,h-asn-otbu hydrochloride
16218498
tert-butyl (2S)-2,4-diamino-4-oxobutanoate;hydrochloride
CC(C)(C)OC(=O)C(CC(=O)N)N.Cl
63094-81-5
C8H17ClN2O3
224.685
MFCD00039079
RXNKCUXXNGWROA-JEDNCBNOSA-N
h-asn-otbu hcl,l-asparagine tert-butyl ester hydrochloride,h-asn-otbu.hcl,tert-butyl l-asparaginate-hydrogen chloride 1/1,l-asparagine, 1,1-dimethylethyl ester, monohydrochloride,tert-butyl 2s-2-amino-3-carbamoylpropanoate hydrochloride,h-asn-otbucl,h-asn-otbu?cl,h-asn-otbuhcl,h-asn-otbu hydrochloride
16218498
tert-butyl (2S)-2,4-diamino-4-oxobutanoate;hydrochloride
CC(C)(C)OC(=O)C(CC(=O)N)N.Cl
Beilstein 6585123
Formula Weight 224.69
Physical Form Powder
Percent Purity 95%
Color White
Quantity 250mg
Chemical Name or Material L-Asparagine tert-butyl ester hydrochloride