Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride, 98%

Click to view available options
Quantity:
250mg
1g
5g
94347-11-2
C15H22ClNO4
315.794
MFCD00190761
YSDLFDMXCRFHTQ-YDALLXLXSA-N
s-4-benzyl 1-tert-butyl 2-aminosuccinate hydrochloride,h-asp obzl-otbu.hcl,4-benzyl 1-tert-butyl 2s-2-aminobutanedioate hydrochloride,c15h21no4.hcl,h-asp obzl-otbu hcl,h-asp obzl-otbu hydrochloride,s-4-benzyl 1-tert-butyl 2-aminosuccinate hcl,l-aspartic acid beta-benzyl ester alfa-t-butyl ester hcl,4-benzyl 1-tert-butyl s-2-aminosuccinate hydrochloride,4-benzyl 1-tert-butyl l-aspartate-hydrogen chloride 1/1
56777343
4-O-benzyl 1-O-tert-butyl (2S)-2-aminobutanedioate;hydrochloride
CC(C)(C)OC(=O)C(CC(=O)OCC1=CC=CC=C1)N.Cl
94347-11-2
C15H22ClNO4
315.794
MFCD00190761
YSDLFDMXCRFHTQ-YDALLXLXSA-N
s-4-benzyl 1-tert-butyl 2-aminosuccinate hydrochloride,h-asp obzl-otbu.hcl,4-benzyl 1-tert-butyl 2s-2-aminobutanedioate hydrochloride,c15h21no4.hcl,h-asp obzl-otbu hcl,h-asp obzl-otbu hydrochloride,s-4-benzyl 1-tert-butyl 2-aminosuccinate hcl,l-aspartic acid beta-benzyl ester alfa-t-butyl ester hcl,4-benzyl 1-tert-butyl s-2-aminosuccinate hydrochloride,4-benzyl 1-tert-butyl l-aspartate-hydrogen chloride 1/1
56777343
4-O-benzyl 1-O-tert-butyl (2S)-2-aminobutanedioate;hydrochloride
CC(C)(C)OC(=O)C(CC(=O)OCC1=CC=CC=C1)N.Cl
Beilstein 2538326
Formula Weight 315.79
Physical Form Solid
Percent Purity 98%
Color White
Quantity 250mg
Chemical Name or Material L-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride

storageNote1 : 2° to 8°C