Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Aspartic Acid, 98.5%, Spectrum™

Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
Poly Pail
56-84-8
C4H7NO4
133.10
CKLJMWTZIZZHCS-UHFFFAOYNA-N
2-aminobutanedioic acid
NC(CC(O)=O)C(O)=O
56-84-8
C4H7NO4
133.10
CKLJMWTZIZZHCS-UHFFFAOYNA-N
2-aminobutanedioic acid
NC(CC(O)=O)C(O)=O

AS125, 56-84-8, C4H7NO4

Spectrum™ L-Aspartic Acid is a nonessential amino acid. Aspartic acid is used as a component of parenteral and enteral nutrition. In pharmaceutical agents aspartic acid is used as an ammoniac detoxicating agent, hepar function accelerator and fatigue refresher. Aspartic acid is also widely utilized for mineral supplementation in the form of its potassium, magnesium or calcium salts. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
CAS Max % 1
Heavy Metals 10 ppm
Ignition Residue 0.001
Lead (Pb) 5 ppm
Loss on Drying 0.002
Packaging Poly Pail
Specific Rotation +24.5° to +26.0°
Percent Purity 98.5%
Quantity 1 kg