Learn More
L-Aspartic acid, 99%, low metals content, Thermo Scientific Chemicals
Description
L-Aspartic acid is used as a component of parenteral and enteral nutrition and as a pharmaceutical ingredient. it is used for cell culture and in manufacturing processes. It is widely utilized for mineral supplementation in the salt form.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >300°C (decomposition) |
Beilstein | 1723530 |
Merck Index | 14,840 |
Solubility Information | Soluble in water(4.5g/L). |
Formula Weight | 133.1 |
Percent Purity | 99% |
Density | 1.66 g/mL |
Quantity | 500 g |
Odor | Odorless |
Physical Form | Powder |
Show More |
Safety and Handling
EINECSNumber | 200-291-6 |
RTECSNumber | CI9098500 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.