Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Aspartic acid dibenzyl ester hydrochloride, 97%

Click to view available options
Quantity:
5g
25g
100g
6327-59-9
C18H20ClNO4
349.811
MFCD01075101
ILBZEWOOBCRRAP-PKLMIRHRSA-N
h-d-asp obzl-obzl.hcl,h-asp obzl-obzl hcl,h-asp obzl-obzl.hcl,1,4-dibenzyl 2r-2-aminobutanedioate hydrochloride
71463668
dibenzyl (2R)-2-aminobutanedioate;hydrochloride
C1=CC=C(C=C1)COC(=O)CC(C(=O)OCC2=CC=CC=C2)N.Cl
6327-59-9
C18H20ClNO4
349.811
MFCD01075101
ILBZEWOOBCRRAP-PKLMIRHRSA-N
h-d-asp obzl-obzl.hcl,h-asp obzl-obzl hcl,h-asp obzl-obzl.hcl,1,4-dibenzyl 2r-2-aminobutanedioate hydrochloride
71463668
dibenzyl (2R)-2-aminobutanedioate;hydrochloride
C1=CC=C(C=C1)COC(=O)CC(C(=O)OCC2=CC=CC=C2)N.Cl
Beilstein 3921431
Formula Weight 349.81
Physical Form Solid
Percent Purity 97%
Quantity 25g
Chemical Name or Material L-Aspartic acid dibenzyl ester hydrochloride

storageNote1 : 2° to 8°C