Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Aspartic acid dibenzyl ester p-toluenesulfonate, 95%

Click to view available options
Quantity:
25g
100g
2886-33-1
C25H27NO7S
485.551
MFCD00065188
HLMUYZYLPUHSNV-NTISSMGPSA-N
h-asp obzl-obzl.tosoh,h-asp obzl-obzl tosoh,l-aspartic acid dibenzyl ester tosylate,s-dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate,l-aspartic acid dibenzyl ester p-toluenesulfonate salt,l-aspartic acid dibenzyl ester 4-toluenesulfonate,h-asp obzl-obzl inverted exclamation mark currencytosoh,h-asp obzl-obzl.tos,l-aspartic acid dibenzyl ester p-toluenesulfonate,1,4-dibenzyl 2s-2-aminobutanedioate; para-toluene sulfonate
2808531
dibenzyl (2S)-2-aminobutanedioate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CC(C(=O)OCC2=CC=CC=C2)N
2886-33-1
C25H27NO7S
485.551
MFCD00065188
HLMUYZYLPUHSNV-NTISSMGPSA-N
h-asp obzl-obzl.tosoh,h-asp obzl-obzl tosoh,l-aspartic acid dibenzyl ester tosylate,s-dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate,l-aspartic acid dibenzyl ester p-toluenesulfonate salt,l-aspartic acid dibenzyl ester 4-toluenesulfonate,h-asp obzl-obzl inverted exclamation mark currencytosoh,h-asp obzl-obzl.tos,l-aspartic acid dibenzyl ester p-toluenesulfonate,1,4-dibenzyl 2s-2-aminobutanedioate; para-toluene sulfonate
2808531
dibenzyl (2S)-2-aminobutanedioate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CC(C(=O)OCC2=CC=CC=C2)N
Beilstein 3647510
Formula Weight 485.55
Physical Form Powder
Percent Purity 95%
Color Brown
Melting Point 157°C to 160°C
Quantity 100g
Chemical Name or Material L-Aspartic acid dibenzyl ester p-toluenesulfonate

storageNote1 : 2° to 8°C