Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Aspartic Acid, FCC, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
100 g
1 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
56-84-8
C4H7NO4
133.10
CKLJMWTZIZZHCS-UHFFFAOYNA-N
2-aminobutanedioic acid
NC(CC(O)=O)C(O)=O
56-84-8
C4H7NO4
133.10
CKLJMWTZIZZHCS-UHFFFAOYNA-N
2-aminobutanedioic acid
NC(CC(O)=O)C(O)=O

A1376, 56-84-8, C4H7NO4

Spectrum™ L-Aspartic Acid, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Ignition Residue 0.001
Lead (Pb) 5 mg/kg
Loss on Drying 0.0025
Packaging Poly Pail
Specific Rotation +24.5° to +26.0°
Percent Purity 98.5 to 101.5%
Quantity 1 kg