Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Aspartic Acid Sodium Salt, Monohydrate, 98-101%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
323194-76-9
C4H8NNaO5
173.10
PPTHNBYUFXSJPS-UHFFFAOYNA-M
sodium 2-amino-3-carboxypropanoate hydrate
O.[Na+].NC(CC(O)=O)C([O-])=O
323194-76-9
C4H8NNaO5
173.10
PPTHNBYUFXSJPS-UHFFFAOYNA-M
sodium 2-amino-3-carboxypropanoate hydrate
O.[Na+].NC(CC(O)=O)C([O-])=O

A1375, 323194-76-9, C4H6NO4Na•H2O

Spectrum™ L-Aspartic Acid Sodium Salt, Monohydrate
CAS Max % 1
Ammonium 0.0004
Arsenic (As) 0.0001
Percent Purity 98 to 101%
Chloride 0.0004
Heavy Metals (as Pb) 0.001
Loss on Drying 0.003
Packaging Poly Bottle
Specific Rotation +19.0° to +21.0°
Sulfate 0.05
Quantity 100 g
Show More Show Less