Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ L-Aspartic Acid Sodium Salt Monohydrate, ≥98% (dry basis), Thermo Scientific™

Click to view available options
Quantity:
1kg
10kg
Packaging:
Plastic bottle
323194-76-9
C4H8NNaO5
173.1
MFCD00152960
PPTHNBYUFXSJPS-JIZZDEOASA-M
sodium l-aspartate,l-aspartic acid sodium salt monohydrate,sodium aspartate monohydrate,sodium l-aspartate monohydrate,s-2-aminobutanedioic acid sodium salt,l-aspartic acid monosodium salt monohydrate,l-aspartic acid, monosodium salt, monohydrate,sodium +-aspartate hydrate,l-aspartic acid sodium salt monohydrate tlc,l-aspartic acid sodium salt monohydrate nt
23679051
sodium;(2S)-2-amino-4-hydroxy-4-oxobutanoate;hydrate
C(C(C(=O)[O-])N)C(=O)O.O.[Na+]
323194-76-9
C4H8NNaO5
173.1
MFCD00152960
PPTHNBYUFXSJPS-JIZZDEOASA-M
sodium l-aspartate,l-aspartic acid sodium salt monohydrate,sodium aspartate monohydrate,sodium l-aspartate monohydrate,s-2-aminobutanedioic acid sodium salt,l-aspartic acid monosodium salt monohydrate,l-aspartic acid, monosodium salt, monohydrate,sodium +-aspartate hydrate,l-aspartic acid sodium salt monohydrate tlc,l-aspartic acid sodium salt monohydrate nt
23679051
sodium;(2S)-2-amino-4-hydroxy-4-oxobutanoate;hydrate
C(C(C(=O)[O-])N)C(=O)O.O.[Na+]
Linear Formula NH2CH(COOH)CH2COONa·H2O
Formula Weight 173.10
Physical Form Crystalline Powder
Packaging Plastic bottle
Quantity 1kg
Chemical Name or Material L to Aspartic Acid Sodium Salt Monohydrate