Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-beta-Homoglutamic acid hydrochloride, 97%

Click to view available options
Quantity:
250mg
1g
61884-74-0
C6H12ClNO4
197.615
MFCD01862872
XMUKSNPNAOIQKU-WCCKRBBISA-N
s-3-aminohexanedioic acid hydrochloride,l-beta-homoglutamic acid hydrochloride,h-beta-hoglu-oh.hcl,3s-3-aminohexanedioic acid hydrochloride,h-beta-homoglu-ohcl,h-hoglu-oh.hcl,h-?-hoglu-oh.hcl,h-beta-homo-glu-oh hcl,l-beta-homoglutamic acidhcl,l-beta-homoglutamic acid-hcl
2761515
(3S)-3-aminohexanedioic acid;hydrochloride
C(CC(=O)O)C(CC(=O)O)N.Cl
61884-74-0
C6H12ClNO4
197.615
MFCD01862872
XMUKSNPNAOIQKU-WCCKRBBISA-N
s-3-aminohexanedioic acid hydrochloride,l-beta-homoglutamic acid hydrochloride,h-beta-hoglu-oh.hcl,3s-3-aminohexanedioic acid hydrochloride,h-beta-homoglu-ohcl,h-hoglu-oh.hcl,h-?-hoglu-oh.hcl,h-beta-homo-glu-oh hcl,l-beta-homoglutamic acidhcl,l-beta-homoglutamic acid-hcl
2761515
(3S)-3-aminohexanedioic acid;hydrochloride
C(CC(=O)O)C(CC(=O)O)N.Cl
Beilstein 6584879
Formula Weight 197.62
Physical Form Solid
Percent Purity 97%
Color White
Quantity 250mg
Chemical Name or Material L-beta-Homoglutamic acid hydrochloride

storageNote1 : 2° to 8°C