Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-beta-Homoleucine hydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
96386-92-4
C7H16ClNO2
181.66
MFCD01862871
NXVYPYHWONGEFQ-RGMNGODLSA-N
l-beta-homoleucine hydrochloride,s-3-amino-5-methylhexanoic acid hydrochloride,h-beta-holeu-oh.hcl,3s-3-amino-5-methylhexanoic acid hydrochloride,h-?-holeu-oh.hcl,hexanoic acid, 3-amino-5-methyl-, hydrochloride, 3s,l-beta-homoleucine-hcl,l-beta-homoleucinehcl,h-b-holeu-oh hcl,h-,a-holeu-oh.hcl
2761524
(3S)-3-amino-5-methylhexanoic acid;hydrochloride
CC(C)CC(CC(=O)O)N.Cl
96386-92-4
C7H16ClNO2
181.66
MFCD01862871
NXVYPYHWONGEFQ-RGMNGODLSA-N
l-beta-homoleucine hydrochloride,s-3-amino-5-methylhexanoic acid hydrochloride,h-beta-holeu-oh.hcl,3s-3-amino-5-methylhexanoic acid hydrochloride,h-?-holeu-oh.hcl,hexanoic acid, 3-amino-5-methyl-, hydrochloride, 3s,l-beta-homoleucine-hcl,l-beta-homoleucinehcl,h-b-holeu-oh hcl,h-,a-holeu-oh.hcl
2761524
(3S)-3-amino-5-methylhexanoic acid;hydrochloride
CC(C)CC(CC(=O)O)N.Cl
Beilstein 8073252
Formula Weight 181.66
Physical Form Powder
Percent Purity 95%
Color Brown to White
Quantity 250mg
Chemical Name or Material L-beta-Homoleucine hydrochloride

storageNote1 : 2° to 8°C