Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-beta-Homoproline hydrochloride, 98%

Click to view available options
Quantity:
250mg
1g
53912-85-9
C6H12ClNO2
165.617
MFCD07363484
VQDACVOAOJQTPR-JEDNCBNOSA-N
s-2-pyrrolidin-2-yl acetic acid hydrochloride,l-beta-homoproline hydrochloride,l-beta-homoproline hcl,2-2s-pyrrolidin-2-yl acetic acid hydrochloride,2-pyrrolidineacetic acid, hydrochloride, 2s,s-2-2-pyrrolidinyl acetic acid hydrochloride,s-3-pyrrolidinecarboxylic acid hydrochloride,2s-pyrrolidin-2-ylacetic acid hydrochloride,l-beta-homoproline-hcl,s-3-pyrrol-idinecarboxylic acid hydrochloride
2761540
2-[(2S)-pyrrolidin-2-yl]acetic acid;hydrochloride
C1CC(NC1)CC(=O)O.Cl
53912-85-9
C6H12ClNO2
165.617
MFCD07363484
VQDACVOAOJQTPR-JEDNCBNOSA-N
s-2-pyrrolidin-2-yl acetic acid hydrochloride,l-beta-homoproline hydrochloride,l-beta-homoproline hcl,2-2s-pyrrolidin-2-yl acetic acid hydrochloride,2-pyrrolidineacetic acid, hydrochloride, 2s,s-2-2-pyrrolidinyl acetic acid hydrochloride,s-3-pyrrolidinecarboxylic acid hydrochloride,2s-pyrrolidin-2-ylacetic acid hydrochloride,l-beta-homoproline-hcl,s-3-pyrrol-idinecarboxylic acid hydrochloride
2761540
2-[(2S)-pyrrolidin-2-yl]acetic acid;hydrochloride
C1CC(NC1)CC(=O)O.Cl
Beilstein 471672
Formula Weight 165.62
Physical Form Powder
Percent Purity 98%
Color White
Quantity 1g
Sensitivity Hygroscopic
Chemical Name or Material L-beta-Homoproline hydrochloride