Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-(-)-Bornyl acetate, 95%

Click to view available options
Quantity:
25g
100g
500g
5655-61-8
C12H20O2
196.29
MFCD00135942
KGEKLUUHTZCSIP-WFCWDVHWSA-N
bornyl acetate,-bornyl acetate
44630108
[(1R,3S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate
CC(=O)OC1CC2CCC1(C2(C)C)C
5655-61-8
C12H20O2
196.29
MFCD00135942
KGEKLUUHTZCSIP-WFCWDVHWSA-N
bornyl acetate,-bornyl acetate
44630108
[(1R,3S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate
CC(=O)OC1CC2CCC1(C2(C)C)C
Boiling Point 223°C to 224°C
Flash Point 84°C (184°F)
Quantity 100g
Beilstein 2502036
Merck Index 14,1339
Formula Weight 196.29
Percent Purity 95%
Chemical Name or Material L-(-)-Bornyl acetate

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA